missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAPSN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115824
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Rapsyn.
This gene encodes a member of a family of proteins that are receptor associated proteins of the synapse. The encoded protein contains a conserved cAMP-dependent protein kinase phosphorylation site, and plays a critical role in clustering and anchoring nicotinic acetylcholine receptors at synaptic sites by linking the receptors to the underlying postsynaptic cytoskeleton, possibly by direct association with actin or spectrin. Mutations in this gene may play a role in postsynaptic congenital myasthenic syndromes. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| RAPSN | |
| Polyclonal | |
| Unconjugated | |
| RAPSN | |
| 43 kD receptor-associated protein of the synapse; 43 kda postsynaptic protein; 43 kDa receptor-associated protein of the synapse; 43kDa; 43kDa acetylcholine receptor-associated protein; acetylcholine receptor-associated 43 kda protein; AChR associated protein at synapses; acetylcholine receptor associated protein at synapses; 43kDa protein; likely to cluster nicotinic AChRs; CMS11; CMS1D; CMS1E; CMS4C; FADS; MGC3597; Nraps; Raps; Rapsn; rapsyn; receptor associated protein of the synapse; receptor-associated protein of the synapse; receptor-associated protein of the synapse, 43 kDa; receptor-associated protein of the synapse, 43kD; RING finger protein 205; RNF205; twitch once; two | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19400, 362161, 5913 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P12672, Q13702 | |
| RAPSN | |
| A synthesized peptide derived from human RAPSN(Accession Q13702), corresponding to amino acid residues E372-V412. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction