missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Rappaport Vassiliadis Enrichment Broth
Description
Rappaport Vassiliadis Enrichment Broth is used to selectively enrich Salmonella species from food and environmental samples by inhibiting competing bacteria and promoting Salmonella growth.
Specifications
Specifications
| Color | Pearl Gentian Blue, Transparent |
| Content And Storage | 2°C to 25°C |
| Format | Poured Tube |
| Manufacturing Origin | Germany |
| Product Type | Microbiology Media |
| Fill Weight | 9.5 to 10.5 g |
| Packaging Type | 50 Tubes/Box |
| Quantity | 50/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?