missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ RapidHIT™ ID Case

Product Code. 30535786 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535786 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30535786 Supplier Applied Biosystems™ Supplier No. A40004464
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The RapidHIT ID Case is a ruggedized, reusable transport case that delivers comprehensive protection and operational flexibility for the RapidHIT ID System in field environments.

The RapidHIT ID Case is a ruggedized, reusable transport case that delivers comprehensive protection and operational flexibility for the RapidHIT ID System in field environments. The case allows users to transform any location into a mobile forensic laboratory, maintaining full RapidDNA capabilities, regardless of location.

The RapidHIT ID Case can be used to store the following components:

  • RapidHIT ID Instrument
  • RapidHIT ID Instrument power cord
  • Drip tray
  • Laptop computer (up to 15 inches)
  • Laptop power cord
  • Ethernet cord (up to 6 feet)
  • Barcode scanner

Engineered to meet ISTA 3B standards, this case ensures uncompromising protection against the shocks and vibrations encountered during standard vehicle transport and commercial freight shipping*. Weather-resistant seals provide a barrier against moisture, dust, and other environmental contaminants, ensuring instrument protection during transportation and extended storage periods.

Features of the RapidHIT ID Case include:

  • Retractable handle
  • ID plate
  • Front label indicators
  • 'This side up' labelling
  • Stainless steel carrying handles
  • Stainless steel latches
  • Airtight certified breather valve
  • Injection molded edge casters
  • Foam plugs

Beyond transportation, the case can convert into a fully integrated workstation upon arrival at your destination. This allows for efficient deployment in space-constrained environments without sacrificing functionality. The integrated design streamlines setup procedures, reduces deployment time, and ensures consistent performance regardless of location – making it an essential component for mobile operation where reliability and efficiency are critical.

For additional recommendations on best practices for use of your RapidHIT ID System in a mobile environment, please refer to the below. For specific instruction on use of the case, refer to the associated below.

*For freight shipping, it is recommended to secure the case to a pallet.

TRUSTED_SUSTAINABILITY

Specifications

Software Type Fragment Analysis
Starting Material DNA
Format Case
For Use With (Equipment) RapidHIT ID System
Height (English) Exterior 29.308 in.
Height (Metric) Exterior 74.442 cm
Length (English) Exterior 25.486 in.
Length (English) Gel 64.734 cm
Length (Metric) Exterior 25.486 in.
Length (Metric) Gel 64.734 cm
Product Type Transport Case
Quantity 1 Ea.
Volume (English) Buffer 17959 in.⊃3
Volume (Metric) Buffer 45615 cm³
Width (English) Exterior 24.044 in.
Width (English) Gel 61.072 cm
Width (Metric) Exterior 24.044 in.
Width (Metric) Gel 61.072 cm
Show More Show Less

For Criminal Justice and Forensic Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.