missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapidFinder™ Turkey ID Kit

Product Code. 17595566
Change view
Click to view available options
Quantity:
48 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17595566 48 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17595566 Supplier Thermo Scientific™ Supplier No. A24394
  • ONLINE EXCLUSIVE PRICE
    £733.00
    £576.20 / Pack of 1
    Save £156.80
    21% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The RapidFinder™ Turkey ID Kit enables the simple and reliable detection of turkey in food and feed samples. The method works by detection of turkey, or Meleagris gallopavo, DNA by real-time PCR.

  • The RapidFinder™ Turkey ID Kit enables the simple and reliable detection of turkey in food and feed samples
  • The method works by detection of turkey, or Meleagris gallopavo, DNA by real-time PCR
  • When used with the GMO Extraction Kit (4466336) for sample preparation, the RapidFinder™ Turkey ID Kit detects both raw and processed turkey and provides sensitivity down to 0.01% turkey DNA
  • The included positive control, which comprises 0.1% turkey DNA, can be used as a reference to set the threshold of detection of the assay
  • Assay is compatible with Applied Biosystems™ StepOne™, StepOnePlus™, 7500, 7500 Fast, and 7300 real-time PCR systems
  • Simple-real-time PCR based assay, no electrophoresis needed
  • Fast-from sample to result in the same day
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 1 tube each of:
Turkey Master Mix (store at -20°C)
General Master Mix (store at -20°C upon receipt; 2-8°C after initial use. Store protected from light)
Positive Control (store at -20°C)
Detection Method Primer-probe
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) Applied Biosystems™ QuantStudio™ 5 Real‑Time PCR Instrument, Applied Biosystems™ 7500 Fast Real‑Time PCR Instrument
Kit Type Turkey ID Kit
Product Line RapidFinder
Quantity 48 reactions
Species Turkey
For Use With (Application) Food and Beverage
No. of Reactions 48 reactions
Sample Type Food & Environmental Samples
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.