missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SureTect™ Salmonella species, Typhimurium and Enteritidis Multiplex PCR Assay

Product Code. 17839142 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Product Code. 17839142 Supplier Thermo Scientific™ Supplier No. A56846

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rapidly detect and differentiate Salmonella species, Salmonella ser. Typhimurium, and Salmonella ser. Enteritidis in a single test with Thermo Scientific™ SureTect™ Salmonella Species, Typhimurium and Enteritidis Multiplex PCR Assay.

Enhance the detection and differentiation of various Salmonella species, including Salmonella ser. Typhimurium and Salmonella ser. Enteritidis, with the Thermo Scientific™ SureTect™ Salmonella Species, Typhimurium and Enteritidis Multiplex PCR Assay. This advanced kit enables you to rapidly identify and distinguish these Salmonella strains in a single test.

With this kit, you can confidently ensure accuracy and expedite product release, as it provides results in as little as 16 hours. The workflows for testing poultry and pork meats, as well as production environment samples, have been validated, ensuring reliable and consistent outcomes.

To further streamline your testing process, the SureTect™ kit is compatible with the Applied Biosystems™ 7500 Fast Real-Time PCR System or Applied Biosystems™ QuantStudio™ 5 Food Safety Real-Time PCR System. These systems offer high-performance capabilities and are designed specifically for food safety applications.

Increase your workflow efficiency while meeting regulatory requirements to detect the most common Salmonella serovars which cause disease in humans with the SureTect Salmonella Species, Typhimurium and Enteritidis Multiplex PCR Assay.

The new SureTect assay kit incorporates color-coded plastics with orientation markings and breakable strip format, colored reagents and pierceable seals to enable an optimized workflow for maximum laboratory efficiency and productivity.

The real-time PCR kit enables rapid, simultaneous detection and presumptive identification of Salmonella species, Salmonella Typhimurium, and Salmonella Enteritidis in a single well, with AFNOR (ISO 16140-2:2016) and AOAC-RI validated workflows on a range of matrices including raw and ready-to-eat poultry and pork meat and food production environment samples.

Benefit from:

  • Faster decisions -  Earlier product release (negative results) and quicker interventions (positive results) due to simultaneous detection (three tests in one) and a reduced time-to-result compared with culture methods.
  • Cost reductions - Up to 50% consumable cost savings compared with running individual PCR tests for Salmonella species, Salmonella Typhimurium, and Salmonella Enteritidis.
  • Increased efficiency -  Save valuable technician time with single, non-proprietary enrichment, ready-to-use reagents, simple sample preparation, and simultaneous identification.

The RapidFInder System is an optimized solution combining speed and performance in an easy-to-use, cost-effective platform—giving you results you can trust.


Simple

  • Easy-to-use with single enrichment step and pre-dispensed reagents
  • Target-specific colored assay plastics
  • Clearly oriented lysis and assay trays
  • Color indicator in lysis reagent reducing possibility for human error

Fast and Flexible

  • Option to automate the lysis and PCR sample preparation
  • Optimized workflow delivers on-screen results in as little as 16 hours
  • Maximum flexibility for operational efficiency
  • Test up to 96 samples in each run
  • Choice of pierceable seals or caps

 Smart

  • Intuitive, user-friendly software delivers reliable results, with no subjective interpretation
  • Actionable data supports fast decisions in a complex product supply chain
  • IPC in every assay for complete confidence in results

The SureTect Salmonella species, Typhimurium and Enteritidis Multiplex PCR Assay includes:

  • Lysis Reagent 1 Tubes
  • Proteinase K
  • Salmonella Multiplex PCR tubes
  • Thermo Scientific 96-Well Pierceable Seals
  • PCR Caps
TRUSTED_SUSTAINABILITY

Specifications

Product Line SureTect
Target Organism Class Salmonella species, Typhimurium and Enteritidis Multiplex
For Use With (Application) Food and Beverage
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.