missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapidFinder™ Ruminant ID Kit

Product Code. 15880017
Change view
Click to view available options
Quantity:
48 reactions
Unit Size:
48 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15880017 48 reactions 48 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15880017 Supplier Thermo Scientific™ Supplier No. A24396
  • ONLINE EXCLUSIVE PRICE
    £637.00
    £562.92 / 48 reactions
    Save £74.08
    11% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The RapidFinder™ Ruminant ID Kit enables the simple and reliable detection of ruminant meat in food and feed samples.

  • Enables the simple and reliable detection of ruminant meat in food and feed samples
  • The method works by detection of Bos taurus, Ovis aries, Capra hircus, Cervus elaphus, and Capreolus capreolus DNA, which corresponds to DNA from beef, sheep, goat, and deer, by real-time PCR
  • Unlike ELISA based methods, when used with the GMO Extraction Kit (4466336) for sample preparation, the RapidFinder™ Ruminant ID Kit detects both raw and processed ruminant meat and provides sensitivity down to 0.01% ruminant DNA
  • The included positive control in the kit, which comprises 0.1% ruminant DNA, can be used as a reference to set the threshold of detection of the assay
  • Simple-real-time PCR based assay, no electrophoresis needed
  • Assay is compatible with Applied Biosystems™ StepOne™, StepOnePlus™, 7500, 7500 Fast, and 7300 real-time PCR systems

Includes

  • Ruminant Master Mix (store at -20°C)
  • General Master Mix (store at 4°C or -20°C)
  • Positive Control (store at -20°C)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 1 tube each of:
Ruminant Master Mix (store at -20°C)
General Master Mix (store at -20°C upon receipt; 2-8°C after initial use. Store protected from light)
Positive Control (store at -20°C)
Detection Method Primer-probe
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) Applied Biosystems™ QuantStudio™ 5 Real‑Time PCR Instrument, Applied Biosystems™ 7500 Fast Real‑Time PCR Instrument
Kit Type Ruminant ID Kit
Product Line RapidFinder
Quantity 48 reactions
Species Ruminant
For Use With (Application) Food and Beverage
No. of Reactions 48 Reactions
Sample Type Food & Environmental Samples
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.