missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapidFinder™ Quant Multi-Animal Set

Product Code. 17515576
Change view
Click to view available options
Quantity:
48 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17515576 48 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17515576 Supplier Thermo Scientific™ Supplier No. A24399

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The RapidFinder™ Quant Multi-Animal Set, when used with one or more of the RapidFinder™ Meat ID kits listed, enables the determination of the percentage of a target meat species with respect to total meat in food and feed samples.

  • The RapidFinder™ Quant Multi-Animal Set, when used with one or more  RapidFinder™ Meat ID kit, enables the determination of the percentage of a target meat species with respect to total meat in food and feed samples
  • Using the included standards, the kit enables quantification of as little as 0.05% target species DNA as compared to other meat DNA in only a few hours
  • The RapidFinder™ Quant Multi-Animal Set is compatible with Applied Biosystems™ StepOne™, StepOnePlus™, 7500, 7500 Fast, and 7300 real-time PCR systems
  • Simple-real-time PCR based assay, no electrophoresis needed
  • Flexible-compatible with any food or feed sample, both raw and processed
  • Reliable-Includes multi-animal DNA quantification standard
  • Fast-from sample to result in the same day

The RapidFinder Quant Multi-Animal Set is designed to be used in combination with one or more of the following meat species-specific ID kits (available separately):

  • RapidFinder™ Beef ID Kit (A24391)
  • RapidFinder™ Pork ID Kit (A24392)
  • RapidFinder™ Equine ID Kit (A15570)
  • RapidFinder™ Poultry ID Kit (A24397)
  • RapidFinder™ Chicken ID Kit (A24393)
  • RapidFinder™ Turkey ID Kit (A24394)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Multi-Animal Master Mix (store at -20°C), General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use. Store protected from light.), Multi-Animal Standard (store at -20°C)
Detection Method Primer-probe
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) Applied Biosystems™ QuantStudio™ 5 Real‑Time PCR Instrument and Applied Biosystems™ 7500 Fast Real‑Time PCR Instrument
Product Line RapidFinder
Quantity 48 reactions
Species Multi-animal
For Use With (Application) Food and Beverage
No. of Reactions 48 reactions
Sample Type Food & Environmental Samples
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.