missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapidFinder™ Pollack ID Kit

Product Code. 17489933
Change view
Click to view available options
Quantity:
48 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17489933 48 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17489933 Supplier Thermo Scientific™ Supplier No. A24404

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Thermo Scientific™ RapidFinder™ Pollack ID Kit enables the simple and reliable detection of pollack (Pollachius pollachius) DNA in food and feed samples.

The RapidFinder™ Pollack ID Kit enables the simple and reliable detection of pollack DNA in food and feed samples. The kit detects samples containing ≥0.01% of pollack DNA.

The RapidFinder™ Pollack ID Kit enables the simple and reliable detection of Pollack DNA in food and feed samples. The method works by detection of pollack (Pollachius pollachius) DNA that is present in food and feed samples by real-time PCR. Unlike ELISA based methods, when used with the GMO Extraction Kit (Cat. No. 4466336) for sample preparation, the RapidFinder™ Pollack ID Kit detects and provides sensitivity down to 0.01% of pollack (Pollachius pollachius) DNA. The included positive control in the kit, which comprises 0.1% Pollack DNA, can be used as a reference to set the threshold of detection of the assay.

  • Simple-real-time PCR based assay, no electrophoresis needed
  • Flexible-pollack DNA detected in any food or feed sample, both raw and processed
  • Reliable-Internal Positive Control (IPC) provides process control check for PCR inhibitors
  • Fast-from sample to result in the same day

The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast real-time PCR systems.

Sample Preparation

DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Part of a Complete Portfolio of Solutions for Fish Species Detection

The RapidFinder™ Pollack ID Kit is just one of our many available kits for the sensitive detection of various fish species in food and feed samples. Other RapidFinder™ Fish species identification kits available include those for the detection of Basa, Gadus, Haddock, Pollock, Tuna, and Specific Tuna.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

Contains 1 tube each of: Pollack Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) Positive Control (store at -20°C) 

Description Pollack ID Kit
Detection Method Primer-probe
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) QuantStudio 5™ and 7500 Fast
Product Line RapidFinder
Product Type RapidFinder™
Quantity 48 reactions
Sensitivity 0.01% of Pollack (Pollachius pollachius) DNA
Species Pollachius pollachius
For Use With (Application) Food and Beverage
Sample Type Food and Environmental Samples
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.