missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapidFinder™ Halal ID Kit

Product Code. 17174205
Click to view available options
Description:
RapidFinder Halal ID Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17174205

Brand: Thermo Scientific™ A47086

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Determine the Halal status of food and feeds using this simple, reliable, sensitive PCR detection method for pork/wild boar DNA.

Simply and reliably detect pork/wild boar DNA in food and feed samples by real-time PCR with the RapidFinder™ Halal ID Kit. Unlike ELISA-based methods which can be less suitable for processed food, when used with the GMO Extraction Kit (Cat no: 4466336) for sample preparation, the RapidFinder Halal ID Kit detects both raw and processed DNA and provides a sensitivity down to 0.0005% (ppm) pork DNA. The included positive control, which represents a 0.1% of pork/wild boar DNA, can be used as a reference to set the threshold of detection of the assay.

Support Halal label claims by testing for the presence of pork/wild boar DNA in food and feed samples to verify if any adulteration or cross-contamination has occurred in purchased ingredients or during manufacture.

  • Simple: real-time PCR based assay, no electrophoresis needed
  • Flexible: pork/wild boar DNA detected in any food or feed sample, both raw and processed
  • Specific: detection of highly specific mitochondrial region from gene 16s common to all pork and wild boar with a limit of detection of 0.0005%
  • Reliable: Internal Positive Control (IPC) provides process-control check for PCR inhibitors
  • Fast: from sample to result in the same day

How the RapidFinder™ Halal ID Kit works
Animal DNA detection and identification is accomplished by real-time PCR using two TaqMan™-MGB probes. The target sequence, detected in the FAM dye channel, specifically detects a mitochondrial DNA sequence specific for Sus scrofa (pork/wild boar). Also included in each reaction mix is an internal positive control detected in the VIC™ dye channel, which is used to verify the absence of PCR inhibitors.

By performing a separate reaction with the included positive control, users are able to verify the system and assay are working correctly. More importantly, it enables easy data interpretation. The positive control represents a 0.1% pork/wild boar DNA, and by comparing sample results to the control results, a threshold of detection is provided.

Sample preparation
DNA sample preparation for the RapidFinder™ Halal ID Kit workflow is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using 10 g of sample, rather than the smaller sample sizes accommodated by most other DNA isolation procedures, minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Quantification
If a positive result is obtained with the RapidFinder™ Halal ID Kit, most labs will go on to quantify the amount of pork/wild boar DNA in the sample. This can be accomplished using the RapidFinder™ Halal ID Kit in conjunction with the RapidFinder™ Quant Multi-Animal Set (Cat. No. A24399).

Part of a suite of solutions for meat species detection
The RapidFinder™ Halal ID Kit is just one of our many kits available for the sensitive detection of various animal species in food and feed samples. Other RapidFinder™ animal species identification kits available include those for the detection of pork, horse, goat, rabbit, water buffalo, duck, beef, horse, sheep, ruminant, chicken, turkey, and poultry.

The RapidFinder™ Animal Species Identification kits are made available through a partnership with Health in Code, S.L.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 1 tube each of: Halal Master Mix (store at -20°C), General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use. Store protected from light), Positive Control (store at -20°C)
Description RapidFinder Halal ID Kit
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
Reaction Speed Fast
Product Type qPCR Kit
For Use With (Application) qPCR
No. of Reactions 48 Reactions
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.