missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapidFinder™ Goat ID Kit

Product Code. 17525576
Click to view available options
Quantity:
48 reactions
This item is not returnable. View return policy

Product Code. 17525576

Brand: Thermo Scientific™ A24407

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Thermo Scientific™ RapidFinder™ Goat ID Kit enables the simple and reliable detection of Goat (Capra aegagrus hircus) DNA in food and feed samples.

The RapidFinder™ Goat ID Kit enables the simple and reliable detection of Goat (Capra aegagrus hircus) DNA in food and feed samples.

The RapidFinder™ Goat ID Kit enables the simple and reliable detection of Goat DNA in food and feed samples. The method works by detection of goat (Capra aegagrus hircus) DNA that is present in food and feed samples by real-time PCR. Unlike ELISA based methods, when used with the GMO Extraction Kit (Cat. No. 4466336) for sample preparation, the RapidFinder™ Goat ID Kit detects and provides sensitivity down to 0.01% of Goat DNA. The included positive control in the kit, which comprises 0.1% Goat DNA, can be used as a reference to set the threshold of detection of the assay.

  • Simple- Real-time PCR based assay, no electrophoresis needed
  • Flexible- Goat DNA detected in any food or feed sample, both raw and processed
  • Reliable- Internal Positive Control (IPC) provides process control check for PCR inhibitors
  • Fast- From sample to result in the same day

The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast Real-Time PCR Systems.

Sample Preparation

DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Quantification

If a positive result is obtained with the RapidFinder™ Goat ID Kit, most labs will go on to quantify the amount of Goat DNA in the sample. This can be accomplished using the RapidFinder™ Goat ID Kit in conjunction with the RapidFinder™ Quant Multi-Animal Set (Cat. No. A24399).

Part of a complete portfolio of Solutions for Meat Species Detection

The RapidFinder™ Goat ID Kit is just one of our many available kits for the sensitive detection of various meat species in food and feed samples. Other RapidFinder™ meat species identification kits available include those for the detection of pork, beef, horse, rabbit, fallow deer, water buffalo, duck, sheep, ruminant, chicken, poultry, and turkey.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 1 tube each of: Goat Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) Positive Control (store at -20°C)
Description Goat ID Kit
Detection Method Primer-probe
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) QuantStudio 5™ and 7500 Fast
Product Line RapidFinder
Product Type RapidFinder™
Quantity 48 reactions
Sensitivity 0.01% of Goat (Capra aegagrus hircus) DNA
Species Capra aegagrus hircus
For Use With (Application) Food and Beverage
Sample Type Food and Environmental Samples
Show More Show Less

The RapidFinder™ Species Identification Kits are made available through a partnership with Health in Code, S.L.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.