missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapidFinder™ Equine ID Kit

Product Code. 17267254
Change view
Click to view available options
Quantity:
48 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17267254 48 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17267254 Supplier Thermo Scientific™ Supplier No. A15570
  • ONLINE EXCLUSIVE PRICE
    £682.35
    £588.72 / Pack of 1
    Save £93.63
    13% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The RapidFinder™ Equine ID Kit enables the simple and reliable detection of horse meat in food and feed samples.

  • Enables the simple and reliable detection of horse meat in food and feed samples
  • Method works by detection of horse (Equus caballus) DNA by real-time PCR
  • Unlike ELISA based methods, the RapidFinder™ Equine ID Kit detects both raw and processed horse meat
  • Provides sensitivity down to 0.01% horse meat DNA when used with the GMO Extraction Kit
  • A positive control is included in the kit (0.1% equine DNA) that can be used as a reference to set the threshold of detection of the assay
  • The assay is compatible with Applied Biosystems™ StepOne™, StepOnePlus™, 7500, 7500 Fast, and 7300 real-time PCR systems

Includes

  • Equine Master Mix: store at -5° to -30°C
  • General Master Mix: store at 2° to 8°C
  • Positive Control: store at -5° to -30°C
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Equine Master Mix (store at -20°C)
General Master Mix (store at -20°C upon receipt; 2-8°C after initial use. Store protected from light)
Positive Control (store at -20°C)
Kit Type Equine ID Kit
Product Line RapidFinder
Quantity 48 reactions
No. of Reactions 48 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.