missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapidFinder™ Duck ID Kit

Product Code. 17835631 Shop All Thermo Scientific Products
Click to view available options
Quantity:
48 reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17835631

Brand: Thermo Scientific™ A24413

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Thermo Scientific™ RapidFinder™ Duck ID Kit enables the simple and reliable detection of duck (genus Anas) DNA in food and feed samples.

The RapidFinder™ Duck ID Kit enables the simple and reliable detection of Duck DNA in food and feed samples.

The RapidFinder™ Duck ID Kit enables the simple and reliable detection of Duck DNA in food and feed samples. The method works by detection of duck (genus Anas) DNA that is present in food and feed samples by real-time PCR. Unlike ELISA based methods, when used with the GMO Extraction Kit (Cat. No. 4466336) for sample preparation, the RapidFinder™ Duck ID Kit detects and provides sensitivity down to 0.01% of duck (genus Anas) DNA. The included positive control in the kit, which comprises 0.1% Duck DNA, can be used as a reference to set the threshold of detection of the assay.

  • Simple- Real-time PCR based assay, no electrophoresis needed
  • Flexible- Duck DNA detected in any food or feed sample, both raw and processed
  • Reliable- Internal Positive Control (IPC) provides process control check for PCR inhibitors
  • Fast- From sample to result in the same day

The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast Real-Time PCR Systems.

Sample Preparation

DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Part of a Complete Portfolio of Solutions for Animal Species Detection

The RapidFinder™ Duck ID Kit is just one of our many available kits for the sensitive detection of various animal species in food and feed samples. Other RapidFinder™ animal species identification kits available include those for the detection of pork, beef, horse, goat, rabbit, fallow deer, sheep, ruminant, chicken, poultry, and turkey.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 1 tube each of: Duck Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) Positive Control (store at -20°C)
Description Duck ID Kit
Detection Method Primer-probe
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) QuantStudio 5™ and 7500 Fast
Product Line RapidFinder
Product Type RapidFinder™
Quantity 48 reactions
Sensitivity 0.01% of Duck (genus Anas) DNA
Species genus Anas
For Use With (Application) Food and Beverage
Sample Type Food and Environmental Samples
Show More Show Less

The RapidFinder™ Species Identification Kits are made available through a partnership with Health in Code, S.L.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.