missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapidFinder™ Basa ID Kit

Product Code. 17499933
Click to view available options
Quantity:
48 reactions
This item is not returnable. View return policy

Product Code. 17499933

Brand: Thermo Scientific™ A24406

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Thermo Scientific™ RapidFinder™ Basa ID Kit enables the simple and reliable detection of basa (Pangasius hypophthalmus) DNA in food and feed samples.

The RapidFinder™ Basa ID Kit enables the simple and reliable detection of Basa DNA in food and feed samples.

The RapidFinder™ Basa ID Kit enables the simple and reliable detection of Basa DNA in food and feed samples. The method works by detection of basa (Pangasius hypophthalmus) DNA that is present in food and feed samples by real-time PCR. Unlike ELISA based methods, when used with the GMO Extraction Kit (Cat. No. 4466336) for sample preparation, the RapidFinder™ Basa ID Kit detects and provides sensitivity down to 0.01% of basa (Pangasius hypophthalmus) DNA. The included positive control in the kit, which comprises 0.1% Basa DNA, can be used as a reference to set the threshold of detection of the assay.

  • Simple- Real-time PCR based assay, no electrophoresis needed
  • Flexible- Basa DNA detected in any food or feed sample, both raw and processed
  • Reliable- Internal Positive Control (IPC) provides process control check for PCR inhibitors
  • Fast- From sample to result in the same day

The assay is compatible with Applied Biosystems™ QuantStudio 5™ and 7500 Fast Real-Time PCR Systems.

Sample Preparation

DNA sample preparation is accomplished with the easy-to-use GMO Extraction Kit (Cat. No. 4466336). Using up to 20 g of sample rather than the smaller sample sizes accommodated by most other DNA isolation procedures minimizes sampling errors. The GMO Extraction Kit is compatible with raw and processed meat, as well as composite food samples.

Part of a Complete Portfolio of Solutions for Fish Species Detection

The RapidFinder™ Basa ID Kit is just one of our many available kits for the sensitive detection of various fish species in food and feed samples. Other RapidFinder™ Fish species identification kits available include those for the detection of Gadus, Pollack, Pollock, Haddock, Tuna, and Specific Tuna.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contains 1 tube each of: Basa Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8 °C after initial use) Positive Control (store at -20°C)
Description Basa ID Kit
Detection Method Primer-probe
PCR Method qPCR
Reaction Speed Standard
For Use With (Equipment) QuantStudio 5™ and 7500 Fast
Product Line

RapidFinder™

Product Type RapidFinder™
Quantity 48 reactions
Sensitivity 0.01% of Basa (Pangasius hypophthalmus) DNA
Species Pangasius hypophthalmus
For Use With (Application) Food and Beverage
Sample Type Food and Environmental Samples
Show More Show Less

The RapidFinder™ Species Identification Kits are made available through a partnership with Health in Code, S.L.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.