missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Rapid V. Cholerae O1 Test is an immunochromatographic membrane assay that uses highly sensitive antibodies to detect V. cholerae antigens in stool sample, environmental water or solid food specimens.
- The reagent pad of the test strip contains colored colloidal gold conjugated with monoclonal antibodies against V. cholerae O1 antigens.
- The reaction membrane contains the secondary antibodies for V. cholerae O1 antigens, which are pre-immobilized in the test line.
- This test is only to be used by healthcare professionals. It cannot be used alone as the basis for the diagnosis and exclusion of cases.
- CE-marked
- Accurate result within 10 minutes
- One-step procedure
- No cross-reaction
- For professional use
Specifications
Specifications
| Disease | Cholera |
| Form | Test card |
| Detection Method | Rapid Detection |
| Sample Type | Environmental Sample, Food Sample |
| Species | Vibrio cholerae O1 |
| Reaction Speed | 10 min. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?