missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapID™ STR System

Product Code. 11474139
Change view
Click to view available options
Quantity:
20 Panels/Kit
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474139 20 Panels/Kit Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11474139 Supplier Thermo Scientific™ Supplier No. R8311003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identify streptococci and related genera in just four hours using this rapid identifcation system based on enzyme technology.

Identify streptococci and related genera in just four hours using the Thermo Scientific™ RapID™ STR System, an identification system based on enzyme technology.

The RapID STR System is a qualitative micromethod employing conventional and chromogenic substrates for the identification of medically important Streptococci and related organisms which have been isolated from human clinical specimens.

  • One-step inoculation—decreased prep time for increased productivity
  • Manual microbial identification made easy—no oil, no pipetting, no 24-hr incubation
  • Four-hour incubation*—quicker time-to-result for a faster response
  • Common procedure—lessened materials usage saves time and money
  • Visible color reactions—reduced subjectivity decreases repeat testing
  • Better reporting—more coverage, advanced updates

*Two-hour incubation for RapID SS/u.

A clear plastic tray contains reagent impregnated wells, allowing simultaneous inoculation of each cavity with a predetermined volume of inoculum. A suspension of test organism in RapID Inoculation Fluid is used as the inoculum which rehydrates and initiates test reactions.

After incubation, each cavity is examined for reactivity by noting the development of a color. In some cases, reagents must be added to the cavities to provide a color change. The resulting pattern of positive and negative scores is used as the basis for identification of the test isolate by comparison of test results to reactivity patterns stored in a database, or through the use of a computer-generated Code Compendium (ERIC).

Requires:
RapID Inoculation Fluid (R8325102).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description RapID STR System
CE Marker Yes
Detectable Analytes Streptococci
Packaging Type 1 Box
Quantity 20 Panels/Kit
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H331-Toxic if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H370-Causes damage to organs.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.