missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapID™ Spot Indole Reagent

Product Code. 12853693
Change view
Click to view available options
Quantity:
15 mL/Vial
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12853693 15 mL/Vial 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12853693 Supplier Thermo Scientific™ Supplier No. R8309002
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Complete the Thermo Scientific™ RapID™ System Indole test using the ready-to-use Thermo Scientific RapID Spot Indole Reagent.

Complete the Thermo Scientific™ RapID™ System Indole test using the Thermo Scientific RapID Spot Indole Reagent.

This product is for use with RapID SS/u System (R8311004), RapID ANA II System (R8311002), RapID NF PLUS System (R8311005), RapID ONE System (R8309002), RapID NH System (R8311001).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description RapID Spot Indole Reagent
CE Marker Yes
Detectable Analytes Gram-negative bacilli (Oxidase-negative)
Format Vial
For Use With (Equipment) RapID SS/u System (R8311004), RapID ANA II System (R8311002), RapID NF PLUS System (R8311005), RapID ONE System (R8309002), RapID NH System (R8311001)
Packaging Type 1 Box
Quantity 15 mL/Vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.