missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Rapid DAB Staining Kit (Colorimetric)

Product Code. 18671855 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
1 set
Product Code. Quantity unitSize
18671855 100 Tests 1 set
1 options
This item is not returnable. View return policy

Product Code. 18671855

Brand: Novus Biologicals™ NBP318633

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Assay Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Staining Kit (Colorimetric)
Quantity 100 Tests
For Use With (Application) Immunohistochemistry (IHC)
Target Specificity Rapid DAB
Content And Storage Store at 4°C in the dark.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.