missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RapID™ CB PLUS System

Product Code. 13245619
Change view
Click to view available options
Quantity:
20 Panels/Kit
Unit Size:
Pack of 20
Product Code. Quantity unitSize
13245619 20 Panels/Kit Pack of 20
1 options
This item is not returnable. View return policy

Product Code. 13245619

Brand: Thermo Scientific™ R8311008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identify Corynebacterium and other gram-positive coryneform bacilli in just four hours using this rapid identifcation system based on enzyme technology.

Identify Corynebacterium and other gram-positive coryneform bacilli in just four hours using the Thermo Scientific™ RapID™ CB PLUS System, an identification system based on enzyme technology.

The RapID CB PLUS System is a qualitative micromethod employing conventional and chromogenic substrates for the identification of medically important coryneform bacteria and other irregular Gram-positive bacilli isolated from human clinical specimens.

  • One-step inoculation—decreased prep time for increased productivity
  • Manual microbial identification made easy—no oil, no pipetting, no 24-hr incubation
  • Four-hour incubation*—quicker time-to-result for a faster response
  • Common procedure—lessened materials usage saves time and money
  • Visible color reactions—reduced subjectivity decreases repeat testing
  • Better reporting—more coverage, advanced updates

*Two-hour incubation for RapID SS/u.

A clear plastic tray contains reagent impregnated wells, allowing simultaneuos inoculation of each cavity with a predetermined volume of inoculum. A suspension of test organism in RapID Innoculation Fluid is used as the inoculum which rehydrates and initiates test reactions.

After incubation, each cavity is examined for reactivity by noting the development of a color. In some cases, reagents must be added to the cavities to provide a color change. The resulting pattern of positive and negative scores is used as the basis for identification of the test isolate by comparison of test results to reactivity patterns stored in a databse or through the use of a computer-generated Code Compedium (ERIC).

Requires:
RapID Spot Indole, (R8309002), and RapID Inoculation Fluid, (R8325106)

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description RapID CB PLUS System
CE Marker Yes
Detectable Analytes Corynebacterium, Gram-positiveCoryneform bacilli
Packaging Type 1 Box
Quantity 20 Panels/Kit
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.