Learn More
3M™ Petrifilm™ Rapid Aerobic Count Plate
Feature technology that facilitates colony enumeration in just 24 hours for most food matrices / environmental samples and resists distortion caused by spreader colonies.
£135.00 - £988.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17273754
|
3M™ Petrifilm™
7100039448 |
50 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17283754
|
3M™ Petrifilm™
7100039449 |
500 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Utilises the convenience and efficiency of the 3M™ Petrifilm™ Plate platform.
- Plates are sample-ready, eliminating the time-consuming, cumbersome step of preparing media/agar dishes.
- Can be used with the 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results.
- This plate has been awarded Official Methods of Analysis (OMA) #2015.13.for Selected Food NF VALIDATION 3M 01/17-11/16 for Dairy products by AFNOR Certification
- Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
- Companies worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Environmental Test |