Learn More
3M™ Petrifilm™ Rapid Aerobic Count Plate
Product Code. 17273754
Click to view available options
Quantity:
50
500
Unit Size:
Pack of 50
Pack of 500
Product Code. 17273754
Brand: 3M™ Petrifilm™ 7100039448
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Feature technology that facilitates colony enumeration in just 24 hours for most food matrices / environmental samples and resists distortion caused by spreader colonies.
  • Utilises the convenience and efficiency of the 3M™ Petrifilm™ Plate platform.
  • Plates are sample-ready, eliminating the time-consuming, cumbersome step of preparing media/agar dishes.
  • Can be used with the 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results.
  • This plate has been awarded Official Methods of Analysis (OMA) #2015.13.for Selected Food NF VALIDATION 3M 01/17-11/16 for Dairy products by AFNOR Certification
  • Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
  • Companies worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Test Type Food and Beverage, Environmental Test
Quantity 50