Learn More
Invitrogen™ RAPH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110270
Description
Immunogen sequence: MEQLSDEEID HGAEEDSDKE DQDLDKMFGA WLGELDKLTQ SLDSDKPMEP VKRSPLRQET NMANFSYRFS IYNLNEALNQ GETVDLDALM ADLCSIEQEL SSIGSGNSKR QITETKATQK LPVSRHTLKH GTLKGLSSSS NRIAKPSHAS YSLDDVTAQL EQASLSMDEA AQQSVLEDTK PLVTNQHRRT ASAGTVSDAE VHSISNSSHS SITSAASSMD SLDIDKVTRP QELDLTHQGQ.
This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.
Specifications
| RAPH1 | |
| Polyclonal | |
| Unconjugated | |
| RAPH1 | |
| 9430025M21Rik; ALS2CR18; ALS2CR9; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 18; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 9; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 18 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 9 protein; C730009O10Rik; KIAA1681; Lamellipodin; LPD; PREL2; PREL-2; proline-rich EVH1 ligand 2; Protein RMO1; RalGDS/AF-6; RAPH1; Ras association (RalGDS/AF-6) and pleckstrin homology domains 1; ras-associated and pleckstrin homology domains-containing protein 1; RMO1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 363239, 65059, 77300 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.69 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q70E73 | |
| RAPH1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.