missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAPGEF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101811
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAPGEF5.
Members of the RAS subfamily of GTPases function in signal transduction as GTP/GDP-regulated switches that cycle between inactive GDP- and active GTP-bound states. Guanine nucleotide exchange factors (GEFs), such as Repac, serve as RAS activators by promoting acquisition of GTP to maintain the active GTP-bound state and are the key link between cell surface receptors and RAS activation. Repac, also known as Rap guanine nucleotide exchange factor 5 (RAPGEF5), is a 580 amino acid protein that is expressed in brain and testis, with weaker expression in heart, placenta, lung, pancreas and small intestine. Existing as two alternatively spliced isoforms, the Repac gene is conserved in cow, mouse and rat. The gene that encodes Repac maps to human chromosome 7p15.3, with high expression of this region linking to positive response to erlotinib therapy for those with non-small-cell lung cancer.
Specifications
| RAPGEF5 | |
| Polyclonal | |
| Unconjugated | |
| RAPGEF5 | |
| 4932413M22; C86120; D030051B22Rik; GFR; guanine nucleotide exchange factor; guanine nucleotide exchange factor for Rap1; KIAA0277; mKIAA0277; mmr-gef; Mras regulated guanine nucleotide exchange factor; M-Ras-regulated GEF; M-Ras-regulated Rap GEF; MRGEF; MR-GEF; Rap guanine nucleotide exchange factor (GEF) 5; Rap guanine nucleotide exchange factor 5; RAPGEF5; Related to Epac; REPAC; RGD1562891; rMR-GEF; tcag7.1034 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 217944, 362799, 9771 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P83900, Q8C0Q9, Q92565 | |
| RAPGEF5 | |
| A synthesized peptide derived from human RAPGEF5(Accession Q92565), corresponding to amino acid residues F521-E571. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction