missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAPGEF5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32220
This item is not returnable.
View return policy
Description
RAPGEF5 Polyclonal specifically detects RAPGEF5 in Human samples. It is validated for Western Blot.
Specifications
| RAPGEF5 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| RAPGEF5 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human RAPGEF5. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Monkey (98%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.74mg/mL | |
| Western Blot 1:500-1:3000 | |
| GFRREPAC, Guanine nucleotide exchange factor for Rap1, KIAA0277M-Ras-regulated GEF, M-Ras-regulated Rap GEF, MRGEF, MR-GEFrap guanine nucleotide exchange factor 5, Rap guanine nucleotide exchange factor (GEF) 5, Related to Epac, Repac | |
| Rabbit | |
| 68 kDa | |
| 0.1 mL | |
| Neuroscience | |
| 9771 | |
| Human | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto