missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAPGEF5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38835-25ul
This item is not returnable.
View return policy
Description
RAPGEF5 Polyclonal specifically detects RAPGEF5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RAPGEF5 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q92565 | |
| RAPGEF5 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EEDLALVAITFSGEKHELQPNDLVISKSLEASGRIYVYRKDLADTLNPFAENEESQQRSMRILGMNTWDL | |
| 25 μL | |
| Neuroscience | |
| 9771 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| GFRREPAC, Guanine nucleotide exchange factor for Rap1, KIAA0277M-Ras-regulated GEF, M-Ras-regulated Rap GEF, MRGEF, MR-GEFrap guanine nucleotide exchange factor 5, Rap guanine nucleotide exchange factor (GEF) 5, Related to Epac, Repac | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction