missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RAPGEF3 Recombinant Rabbit Monoclonal Antibody (K01_1Q83)

Product Code. 30828792
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828792 50 μL
30827163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828792 Supplier Invitrogen™ Supplier No. MA565272
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

RAPGEF3 Recombinant Monoclonal Antibody for Western Blot

Gene ontology (GO) annotation includes small GTPase mediated signal transduction. Target protein binds cAMP and activates the Ras superfamily guanine nucleotide binding protein (Rap1A)in a PKA-independent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPGEF3
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1Q83
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene RAPGEF3
Gene Accession No. O95398, Q8VCC8
Gene Alias 2310016P22Rik; 9330170P05Rik; bcm910; CAMP-GEFI; cAMP-regulated guanine nucleotide exchange factor I; cAMP-regulated guanine nucleotide exchange factor I (cAMP-GEFI); CGEF1; Epac; EPAC 1; Epac1; Exchange factor directly activated by cAMP 1; exchange protein directly activated by cAMP 1; HSU79275; LOW QUALITY PROTEIN: rap guanine nucleotide exchange factor 3; MGC21410; Rap guanine nucleotide exchange factor (GEF) 3; Rap guanine nucleotide exchange factor 3; RAP guanine-nucleotide-exchange factor 3; Rap1 guanine-nucleotide-exchange factor directly activated by cAMP; Rapgef3
Gene Symbols RAPGEF3
Host Species Rabbit
Immunogen A synthetic peptide of human Epac1.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10411, 223864
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.