missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAPGEF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572745
This item is not returnable.
View return policy
Description
RAPGEF3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Gene ontology (GO) annotation includes small GTPase mediated signal transduction. Target protein binds cAMP and activates the Ras superfamily guanine nucleotide binding protein (Rap1A)in a PKA-independent manner.
Specifications
| RAPGEF3 | |
| Polyclonal | |
| Unconjugated | |
| RAPGEF3 | |
| 2310016P22Rik; 9330170P05Rik; bcm910; CAMP-GEFI; cAMP-regulated guanine nucleotide exchange factor I; cAMP-regulated guanine nucleotide exchange factor I (cAMP-GEFI); CGEF1; Epac; EPAC 1; Epac1; Exchange factor directly activated by cAMP 1; exchange protein directly activated by cAMP 1; HSU79275; LOW QUALITY PROTEIN: rap guanine nucleotide exchange factor 3; MGC21410; Rap guanine nucleotide exchange factor (GEF) 3; Rap guanine nucleotide exchange factor 3; RAP guanine-nucleotide-exchange factor 3; Rap1 guanine-nucleotide-exchange factor directly activated by cAMP; Rapgef3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10411, 223864, 59326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O95398, Q8VCC8, Q9Z1C8 | |
| RAPGEF3 | |
| EPAC1 antibody for an 18 amino acid synthetic peptide near the amino terminus of human EPAC1. The immunogen is within amino acids 60 - 110 of EPAC1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction