missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAPGEF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559147
This item is not returnable.
View return policy
Description
Immunogen sequence: LAVEDSPALG APRVGALPDV VPEGTLLNMV LRRMHRPRSC SYQLLLEHQR PSCIQGLRWT PLTNSEESLD FSESLEQAST ERVLRAGRQL HRHLLATCPN L Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 77%.
Gene ontology (GO) annotation includes small GTPase mediated signal transduction. Target protein binds cAMP and activates the Ras superfamily guanine nucleotide binding protein (Rap1A)in a PKA-independent manner.
Specifications
| RAPGEF3 | |
| Polyclonal | |
| Unconjugated | |
| RAPGEF3 | |
| 2310016P22Rik; 9330170P05Rik; bcm910; CAMP-GEFI; cAMP-regulated guanine nucleotide exchange factor I; cAMP-regulated guanine nucleotide exchange factor I (cAMP-GEFI); CGEF1; Epac; EPAC 1; Epac1; Exchange factor directly activated by cAMP 1; exchange protein directly activated by cAMP 1; HSU79275; LOW QUALITY PROTEIN: rap guanine nucleotide exchange factor 3; MGC21410; Rap guanine nucleotide exchange factor (GEF) 3; Rap guanine nucleotide exchange factor 3; RAP guanine-nucleotide-exchange factor 3; Rap1 guanine-nucleotide-exchange factor directly activated by cAMP; Rapgef3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10411 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95398 | |
| RAPGEF3 | |
| Recombinant protein corresponding to Human RAPGEF3. Recombinant protein control fragment (Product #RP-97655). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction