missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAPGEF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37986-20ul
This item is not returnable.
View return policy
Description
RAPGEF3 Polyclonal antibody specifically detects RAPGEF3 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| RAPGEF3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| 9330170P05Rik, bcm910, cAMP-GEFI, cAMP-regulated guanine nucleotide exchange factor I, CGEF1, EPAC 1, EPAC1, EPACRAP guanine-nucleotide-exchange factor (GEF) 3, Exchange factor directly activated by cAMP 1, Exchange protein directly activated by cAMP 1, HSU79275, MGC21410, Rap guanine nucleotide exchange factor (GEF) 3, rap guanine nucleotide exchange factor 3, Rap1 guanine-nucleotide-exchange factor directly activated by cAMP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human RAPGEF3 (NP_001092002.1).,, Sequence:, MVLRRMHRPRSCSYQLLLEHQRPSCIQGLRWTPLTNSEESLDFSESLEQASTERVLRAGRQLHRHLLATCPNLIRDRKYHLRLYRQCCSGRELVDGILALGLGVHSRSQVVGICQVLLDEGALCHVKHDWAFQDRDAQFYRFPGPEPEPVGTHEMEEELAEAVALLSQRGPDALLTVALRKPPGQRTDEELDLIFEELLH | |
| 20 μL | |
| Signal Transduction | |
| 10411 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction