missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAPGEF3 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81767CL1
This item is not returnable.
View return policy
Description
RAPGEF3 Polyclonal antibody specifically detects RAPGEF3 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| RAPGEF3 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| 9330170P05Rik, bcm910, cAMP-GEFI, cAMP-regulated guanine nucleotide exchange factor I, CGEF1, EPAC 1, EPAC1, EPACRAP guanine-nucleotide-exchange factor (GEF) 3, Exchange factor directly activated by cAMP 1, Exchange protein directly activated by cAMP 1, HSU79275, MGC21410, Rap guanine nucleotide exchange factor (GEF) 3, rap guanine nucleotide exchange factor 3, Rap1 guanine-nucleotide-exchange factor directly activated by cAMP | |
| RAPGEF3 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human RAPGEF3. The immunogen is located within amino acids 60 - 110 of RAPGEF3. Amino Acid Squence: WTPLTNSEESLDFSESLE | |
| 0.1 mL | |
| Signal Transduction | |
| 10411 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction