missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rapamycin, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
Unit Size:
100mg
1g
200mg
2g
500mg
50mg
5g
Description
Rapamycin (Sirolimus
Specifications
Specifications
| Chemical Name or Material | Rapamycin |
| CAS | 53123-88-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C51H79NO13 |
| Quantity | 1 g |
| Synonym | Sirolimus AY-22989 |
| Solubility Information | DMSO : 125 mg/mL (136.74 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (54.69 mM; Need ultrasonic) |
| SMILES | O=C([C@@]1(O)[C@@H](CC[C@@H](C[C@@H](/C(C)=C/C=C/C=C/[C@H](C[C@@H](C)C([C@@H]([C@@H](/C(C)=C/[C@H]2C)O)OC)=O)C)OC)O1)C)C(N3CCCC[C@H]3C(O[C@@H](CC2=O)[C@@H](C[C@H]4C[C@H]([C@H](O)CC4)OC)C)=O)=O |
| Molecular Weight (g/mol) | 914.17 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction