missing translation for 'onlineSavingsMsg'
Learn More

Rapamycin, MedChemExpress

Product Code. 16404796
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
Unit Size:
100mg
1g
200mg
2g
500mg
50mg
5g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16404796 100 mg 100mg
16451546 500 mg 500mg
16450979 1 g 1g
16440956 200 mg 200mg
16430076 2 g 2g
16420107 50 mg 50mg
16436457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16404796 Supplier Medchem Express Supplier No. HY10219
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Rapamycin (Sirolimus; AY 22989) is a potent and specific mTOR inhibitor with an IC50 of 0.1 nM in HEK293 cells. Rapamycin binds to FKBP12 and specifically acts as an allosteric inhibitor of mTORC1. Rapamycin is an autophagy activator, an immunosuppressant.

Rapamycin (Sirolimus

Specifications

Chemical Name or Material Rapamycin
CAS 53123-88-9
Color White
Purity Grade Notes Research
Molecular Formula C51H79NO13
Quantity 100 mg
Synonym Sirolimus AY-22989
Solubility Information DMSO : 125 mg/mL (136.74 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (54.69 mM; Need ultrasonic)
SMILES O=C([C@@]1(O)[C@@H](CC[C@@H](C[C@@H](/C(C)=C/C=C/C=C/[C@H](C[C@@H](C)C([C@@H]([C@@H](/C(C)=C/[C@H]2C)O)OC)=O)C)OC)O1)C)C(N3CCCC[C@H]3C(O[C@@H](CC2=O)[C@@H](C[C@H]4C[C@H]([C@H](O)CC4)OC)C)=O)=O
Molecular Weight (g/mol) 914.17
Formula Weight 914.17
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.