missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAP80 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557978
This item is not returnable.
View return policy
Description
Immunogen sequence: PVFENVNVKS FDRCTGHSAE HTQCGKPQES TGRGSAFLKA VQGSGDTSRH CLPTLADAKG LQDTGGTVNY FWGIPFCPDG VDPNQYTKVI LCQLE Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.
RAP80 was initially identified as zinc-finger containing nuclear protein that is highly expressed in testis and interacts with the retinoid-related testis-associated receptor (RTR). Later experiments revealed that RAP80 is recruited by the Coiled-coil domain 98 (CCDC98) protein to the breast cancer-1 protein BRCA1, allowing the formation of BRCA1 foci in response to DNA damage caused by ionizing radiation. Both RAP80 and CCDC98 are required for DNA damage resistance, G2-M checkpoint control, and DNA repair. Cells depleted of either RAP80 or CCDC98 exhibited increased sensitivity to ionizing radiation, although not as much as in BRCA1-depleted cells, suggesting that RAP80 and CCDC98 control only part of the DNA damage response role of BRCA1. At least four isoforms of RAP80 are known to exist.
Specifications
| RAP80 | |
| Polyclonal | |
| Unconjugated | |
| UIMC1 | |
| 9430016E08Rik; BRCA1-A complex subunit RAP80; D330018D10Rik; D630032M02Rik; RAP80; Receptor-associated protein 80; retinoid X receptor interacting protein 110; retinoid X receptor-interacting protein 110; RGD1307009; RIP110; Rxrip110; ubiquitin interaction motif containing 1; ubiquitin interaction motif-containing protein 1; Uimc1; X2HRIP110 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51720 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96RL1 | |
| UIMC1 | |
| Recombinant protein corresponding to Human RAP80. Recombinant protein control fragment (Product #RP-96861). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction