missing translation for 'onlineSavingsMsg'
Learn More

RAP80 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18412514 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412514 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412514 Supplier Novus Biologicals Supplier No. NBP176828AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAP80 Polyclonal antibody specifically detects RAP80 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP80
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias BRCA1-A complex subunit RAP80, RAP80X2HRIP110, receptor associated protein 80, Receptor-associated protein 80, retinoid x receptor interacting protein, Retinoid X receptor-interacting protein 110, RXRIP110, ubiquitin interaction motif containing 1, Ubiquitin interaction motif-containing protein 1
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the carboxy terminus of human RAP80. The immunogen is located within amino acids 520 - 570 of RAP80. Amino Acid Squence: TRRQKEAKTKSDSGTA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 51720
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.