missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP74 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£301.00 - £478.00
Specifications
| Antigen | RAP74 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18378213
|
Bio-Techne
NBP3-17976-25UL |
25 μg |
£301.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18300455
|
Novus Biologicals
NBP3-17976-100UL |
100 μg |
£478.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAP74 Polyclonal antibody specifically detects RAP74 in Human samples. It is validated for ImmunofluorescenceSpecifications
| RAP74 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research | |
| PBS (pH 7.2) and 40% Glycerol | |
| 2962 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BTF4, General transcription factor IIF 74 kDa subunit, general transcription factor IIF subunit 1, general transcription factor IIF, polypeptide 1, 74kDa, TF2F1, TFIIF, TFIIF-alpha, Transcription initiation factor IIF subunit alpha, transcription initiation factor RAP74 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence: QEVDYMSDGSSSSQEEPESKAKAPQQEEGPKGVDEQSDSSEESEEEKPPEEDKEEEEEKKAPTPQEKKRRKDSSEESDSSEESDIDSEASSALFMAKKKTPPKRERKPSGGSSRGNSRPGTPSAEGGSTSSTLRAAASKLEQGK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title