missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP74 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35898-20ul
This item is not returnable.
View return policy
Description
RAP74 Polyclonal antibody specifically detects RAP74 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| RAP74 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| BTF4, General transcription factor IIF 74 kDa subunit, general transcription factor IIF subunit 1, general transcription factor IIF, polypeptide 1, 74kDa, TF2F1, TFIIF, TFIIF-alpha, Transcription initiation factor IIF subunit alpha, transcription initiation factor RAP74 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 390-517 of RAP74 (NP_002087.2).,, Sequence:, PSAEGGSTSSTLRAAASKLEQGKRVSEMPAAKRLRLDTGPQSLSGKSTPQPPSGKTTPNSGDVQVTEDAVRRYLTRKPMTTKDLLKKFQTKKTGLSSEQTVNVLAQILKRLNPERKMINDKMHFSLKE | |
| 20 μL | |
| Chromatin Research | |
| 2962 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction