missing translation for 'onlineSavingsMsg'
Learn More

RAP30 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Product Code. 18354614
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354614 0.1 mL 0.1mL
18318993 0.02 mL 0.02mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18354614 Supplier Novus Biologicals Supplier No. NBP3179820.1ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RAP30 Polyclonal antibody specifically detects RAP30 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAP30
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias ATP-dependent helicase GTF2F2, BTF4, General transcription factor IIF 30 kDa subunit, general transcription factor IIF subunit 2, general transcription factor IIF, polypeptide 2, 30kDa, TF2F2, TFIIF, TFIIF-beta, Transcription initiation factor IIF subunit beta, Transcription initiation factor RAP30
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-249 of human GTF2F2 (NP_004119.1).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 2963
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.