missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAP2C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101831
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAP2C.
Small GTP-binding protein which cycles between a GDP-bound inactive and a GTP-bound active form. May play a role in cytoskeletal rearrangements and regulate cell spreading through activation of the effector TNIK. May play a role in SRE-mediated gene transcription.
Specifications
| RAP2C | |
| Polyclonal | |
| Unconjugated | |
| RAP2C | |
| 2010200P20Rik; AI194294; AL022976; RAP2C; RAP2C, member of RAS oncogene family; ras-related protein Rap-2c; small GTPase RAP2C | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 302495, 57826, 72065 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BU31, Q9Y3L5 | |
| RAP2C | |
| A synthesized peptide derived from human RAP2C(Accession Q9Y3L5), corresponding to amino acid residues M96-S146. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction