Learn More
RAP2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530144
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431, H1299, HepG2, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (98%), Xenopus laevis (96%), Pig (100%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This intronless gene belongs to a family of RAS-related genes. The proteins encoded by these genes share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between the RAP and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. Evidence suggests that this protein may be polyisoprenylated and palmitoylated.Specifications
RAP2B | |
Polyclonal | |
Unconjugated | |
RAP2B | |
4021402C18Rik; AA408554; Rap2b; RAP2B, member of RAS oncogene family; ras-related protein Rap-2b; small GTP binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5912, 74012 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.97 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P61225, P61226 | |
RAP2B | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 149 of Human RAP2B | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |