missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAP2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101830
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RAP2B.
This intronless gene belongs to a family of RAS-related genes. The proteins encoded by these genes share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between the RAP and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. Evidence suggests that this protein may be polyisoprenylated and palmitoylated.
Specifications
| RAP2B | |
| Polyclonal | |
| Unconjugated | |
| RAP2B | |
| 4021402C18Rik; AA408554; Rap2b; RAP2B, member of RAS oncogene family; ras-related protein Rap-2b; small GTP binding protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 170923, 5912, 74012 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P61225, P61226, P61227 | |
| RAP2B | |
| A synthesized peptide derived from human RAP2B(Accession P61225), corresponding to amino acid residues R97-A147. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction