missing translation for 'onlineSavingsMsg'
Learn More

Rap2A Antibody, HRP, Novus Biologicals™

Codice prodotto. 18429441
Cambia vista
Click to view available options
Quantity:
0.1 mL
Dimensione della confezione:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
18429441 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18429441 Fornitore Novus Biologicals (Bio-Techne) N. del fornitore NBP224574H

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

Rap2A Polyclonal antibody specifically detects Rap2A in Human, Mouse, Canine, Chicken, Equine, Opossum, Primate, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifica

Antigen Rap2A
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias KREV, K-REV, RAP2, RAP2, member of RAS oncogene family (K-rev), RAP2A, member of RAS oncogene family, ras-related protein Rap-2a, RbBP-30
Host Species Rabbit
Immunogen A portion of amino acids 140-183 of human RAP2A was used as the immunogen for the antibody.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5911
Target Species Human, Mouse, Canine, Chicken, Equine, Opossum, Primate, Xenopus species
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.