missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rap2A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93956-0.1ml
This item is not returnable.
View return policy
Description
Rap2A Polyclonal antibody specifically detects Rap2A in Human, Mouse samples. It is validated for Western Blot
Specifications
| Rap2A | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| KREV, K-REV, RAP2, RAP2, member of RAS oncogene family (K-rev), RAP2A, member of RAS oncogene family, ras-related protein Rap-2a, RbBP-30 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-183 of human RAP2A (NP_066361.1). MREYKVVVLGSGGVGKSALTVQFVTGTFIEKYDPTIEDFYRKEIEVDSSPSVLEILDTAGTEQFASMRDLYIKNGQGFILVYSLVNQQSFQDIKPMRDQIIRVKRYEKVPVILVGNKVDLESEREVSSSEGRALAEEWGCPFMETSAKSKTMVDELFAEIVRQMNYAAQPDKDDPCCSACNIQ | |
| 0.1 mL | |
| Signal Transduction | |
| 5911 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction