missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP1GAP Mouse anti-Human, Clone:2G7B6, Abnova™
Mouse monoclonal antibody raised against recombinant human RAP1GAP.
Brand: Abnova MAB16625.100ug
This item is not returnable.
View return policy
Specifications
| RAP1GAP | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| RAP1GAP | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Flow Cytometry, Western Blot | |
| 2G7B6 | |
| RAP1 GTPase activating protein | |
| In PBS (0.05% sodium azide) | |
| KIAA0474, RAP1GA1, Rap1GAP1, rap1GAPII | |
| Recombinant protein corresponding to amino acid 412-678 of human RAP1GAP from E. coli. | |
| 5909 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction