missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10840-1-AP-20UL
This item is not returnable.
View return policy
Description
RAP1B and RAP1A belong to a superfamily of RAS like small GTP-binding proteins involved in cell signaling.Specifications
| RAP1B | |
| Polyclonal | |
| Unconjugated | |
| RAP1B | |
| GTP binding protein smg p21B, K REV, RAL1B, RAP1B, Ras related protein Rap 1b | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171337, 215449, 5908 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P61224, Q62636, Q99JI6 | |
| Rap1b | |
| RAP1B Fusion Protein Ag1282 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction