missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAP1A/RAP1B Recombinant Rabbit Monoclonal Antibody (24GB1030)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | RAP1A/RAP1B |
|---|---|
| Clone | 24GB1030 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275287
![]() |
Invitrogen™
MA557209 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275551
|
Invitrogen™
MA557210 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RAP1A/RAP1B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
| RAP1A/RAP1B | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| RAP1A | |
| 2810443E11Rik; AI848598; C21KG; G-22K; GTP-binding protein smg p21A; GTP-binding protein smg p21B; GTP-binding protein SMG-P21A; K-REV; KREV1; Krev-1; OK/SW-cl.11; RAL1B; rap 1A; RAP1; RAP1A; RAP-1A; RAP1A, member of RAS oncogene family; Rap1b; RAP1B, member of RAS oncogene family; Ras family small GTP binding protein RAP1B; RAS related protein 1b; RAS-related protein 1a; Ras-related protein Krev-1; Ras-related protein Rap-1A; RAS-related protein RAP1B; ras-related protein Rap-1b; RAS-related protein-1a; small GTP binding protein; SMGP21 | |
| RAP1A, RAP1B | |
| Primary | |
| Affinity chromatography |
| 24GB1030 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P61224, P62834, P62835, P62836, Q62636, Q99JI6 | |
| 109905, 171337, 215449, 295347, 5906, 5908 | |
| A synthesized peptide derived from human RAP1A/RAP1B (aa 100-150). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title