missing translation for 'onlineSavingsMsg'
Learn More

RAP1A Antibody (1D2-1C64), Novus Biologicals™

Mouse Monoclonal Antibody

£159.00

Specifications

Antigen RAP1A
Clone 1D2-1C64
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:1000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:100 - 1:1000
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18793394
View Documents
Novus Biologicals
NBP2-22526-0.025mg
0.025 mg
£159.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RAP1A Monoclonal specifically detects RAP1A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

RAP1A
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cell Cycle and Replication
P62834
5906
Recombinant full-length human Rap1
Primary
Store at -20°C. Avoid freeze-thaw cycles.
21 kDa
1D2-1C64
Western Blot 1:500 - 1:1000, Immunohistochemistry 1:100 - 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunoprecipitation 2 μg, Immunohistochemistry-Paraffin 1:100 - 1:1000
Monoclonal
Purified
RUO
Human, Mouse
C21KG, G-22K, GTP-binding protein smg p21A, KREV-1, KREV1Ras-related protein Krev-1, RAP1, RAP1A, member of RAS oncogene family, ras-related protein Rap-1A, RAS-related protein RAP1A, SMGP21
RAP1A
IgG2a
Protein A purified
This product was made specifically to the RAP1A human protein (P62834). It has not yet been tested for cross reactivity to other related proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.