missing translation for 'onlineSavingsMsg'
Learn More

Ranolazine, MedChemExpress

Product Code. 16451406
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451406 200 mg 200mg
16442086 500 mg 500mg
16408216 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16451406 Supplier Medchem Express Supplier No. HYB0280
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ranolazine (CVT 303) is an anti-angina drug that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine is also a partial fatty acid oxidation (FAO) inhibitor. Antianginal agent.

Ranolazine (CVT 303) is an anti-angina drug that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine is also a partial fatty acid oxidation (FAO) inhibitor. Antianginal agent.

Specifications

Chemical Name or Material Ranolazine
CAS 95635-55-5
Color White
Purity Grade Notes Research
Molecular Formula C24H33N3O4
Quantity 200 mg
Synonym CVT 303 RS 43285-003
Solubility Information DMSO : 125 mg/mL (292.37 mM; Need ultrasonic)
SMILES O=C(NC1=C(C)C=CC=C1C)CN2CCN(CC(O)COC3=CC=CC=C3OC)CC2
Molecular Weight (g/mol) 427.54
Formula Weight 427.54
Percent Purity 99.72%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.