missing translation for 'onlineSavingsMsg'
Learn More

Ranolazine dihydrochloride, MedChemExpress

Product Code. 16441256
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
200mg
500mg
5g
This item is not returnable. View return policy

Product Code. 16441256

Brand: Medchem Express HY17401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ranolazine dihydrochloride (CVT 303 dihydrochloride) is an anti-angina drug that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine dihydrochloride is also a partial fatty acid oxidation inhibitor.

Ranolazine dihydrochloride (CVT 303 dihydrochloride) is an anti-angina drug that achieves its effects by inhibiting the late phase of inward sodium current (INa and IKr with IC50 values of 6 μM and 12 μM, respectively) without affecting heart rate or blood pressure (BP). Ranolazine dihydrochloride is also a partial fatty acid oxidation inhibitor.

Specifications

Chemical Name or Material Ranolazine dihydrochloride
CAS 95635-56-6
Color White
Purity Grade Notes Research
Molecular Formula C24H35Cl2N3O4
Quantity 200 mg
Synonym CVT 303 dihydrochloride RS 43285
Solubility Information DMSO : ≥ 50 mg/mL (99.91 mM) ∣H2O : ≥ 50 mg/mL (99.91 mM)
SMILES O=C(NC1=C(C)C=CC=C1C)CN2CCN(CC(O)COC3=CC=CC=C3OC)CC2.[H]Cl.[H]Cl
Molecular Weight (g/mol) 500.46
Formula Weight 500.46
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.