missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RANK Recombinant Rabbit Monoclonal Antibody (2Z1P2)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557952

 View more versions of this product

Product Code. 30284364

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: IAPPCTSEKH YEHLGRCCNK CEPGKYMSSK CTTTSDSVCL PCGPDEYLDS WNEEDKCLLH KVCDTGKALV AVVAGNSTTP RRCACTAGYH WSQDCECCRR NTECAPGLGA QHPLQLNKDT VCKPCLAGYF SDAFSSTDKC RPWTNCTFLG KRVEHHGTEK SDAVCSSSLP ARKPPNEPHV YLP.

RANK is a member of the tumor necrosis factor receptor (TNFR) family. The full length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RANK
Recombinant Monoclonal
0.96 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
Q9Y6Q6
TNFRSF11A
Recombinant protein (or fragment)
100 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
2Z1P2
Unconjugated
TNFRSF11A
CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11a; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Rabbit
Protein A
RUO
8792
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.