missing translation for 'onlineSavingsMsg'
Learn More

Ranitidine hydrochloride, MedChemExpress

Product Code. 16418216
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16418216

Brand: Medchem Express HYB0281A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ranitidine hydrochloride is a potent, selective and orally active histamine H2-receptor antagonist with an IC50 of 3.3 μM that inhibits gastric secretion. Ranitidine hydrochloride is a weak inhibitor of CYP2C19 and CYP2C9.

Ranitidine hydrochloride is a potent, selective and orally active histamine H2-receptor antagonist with an IC50 of 3.3 μM that inhibits gastric secretion. Ranitidine hydrochloride is a weak inhibitor of CYP2C19 and CYP2C9.

Specifications

Chemical Name or Material Ranitidine hydrochloride
Physical Form Solid
CAS 66357-59-3
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C13H23ClN4O3S
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information H2O : ≥ 50 mg/mL (142.51 mM) ∣DMSO : 50 mg/mL (142.51 mM; Need ultrasonic)
SMILES CN/C(NCCSCC1=CC=C(CN(C)C)O1)=C\[N+]([O-])=O.Cl
Molecular Weight (g/mol) 350.86
Formula Weight 350.86
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.