missing translation for 'onlineSavingsMsg'
Learn More

Ranitidine hydrochloride, MedChemExpress

Product Code. 16452086
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452086 500 mg 500mg
16418216 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452086 Supplier Medchem Express Supplier No. HYB0281A
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ranitidine hydrochloride is a potent, selective and orally active histamine H2-receptor antagonist with an IC50 of 3.3 μM that inhibits gastric secretion. Ranitidine hydrochloride is a weak inhibitor of CYP2C19 and CYP2C9.

Ranitidine hydrochloride is a potent, selective and orally active histamine H2-receptor antagonist with an IC50 of 3.3 μM that inhibits gastric secretion. Ranitidine hydrochloride is a weak inhibitor of CYP2C19 and CYP2C9.

Specifications

Chemical Name or Material Ranitidine hydrochloride
Physical Form Solid
CAS 66357-59-3
Color Off-White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C13H23ClN4O3S
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information H2O : ≥ 50 mg/mL (142.51 mM) ∣DMSO : 50 mg/mL (142.51 mM; Need ultrasonic)
SMILES CN/C(NCCSCC1=CC=C(CN(C)C)O1)=C\[N+]([O-])=O.Cl
Molecular Weight (g/mol) 350.86
Formula Weight 350.86
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.